missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PET Storage Bottles, Square with Cap

Catalog No. 07201603
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
Graduations:
100 to 1000 mL
100 to 500 mL
50 to 125 mL
50 to 250 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Graduations
07201603 1 L 100 to 1000 mL
07201600 125 mL 50 to 125 mL
07201601 250 mL 50 to 250 mL
07201602 500 mL 100 to 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07201603 Supplier Corning™ Supplier No. 431533
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage

Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage. PET resin is lightweight, break-resistant and transparent as glass. Bottles use a linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.

Features:

  • Good for storage of media, buffers and other aqueous solutions
  • Wide neck provides easier access for liquid transfer
  • Screened white enamel graduations are easier to read than molded graduations
  • Sterilized to SAL 10−6
  • Large white marking spot for easier identification and easy opening linear tear packaging
  • Individually bagged
  • Validated USP Class VI, noncytotoxic and nonpyrogenic


  •  

Specifications

Capacity 33.8 oz., 1000 mL
Capacity (Metric) 1 L
Color Orange
Diameter (English) 3.6 in.
Diameter (English) Inner Neck 1.4 in.
Diameter (Metric) Inner Neck 37 mm
For Use With (Application) Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage
Material PET
No. per Case 24/Cs.
No. per Pack 12/Pk.
Quantity 12 Pk.
Sterility Sterile
Thread Size (Metric) GL 45
Closure Color Orange
Graduations 100 to 1000 mL
Graduated Yes
Height (Metric) 216.5 mm
Height (English) 8.5 in.
Closure Material HDPE
Closure Size GL 45
Diameter (Metric) 92 mm
Closure Type Plug Seal
Capacity (English) 33.8 oz.
Shape Square
Certifications/Compliance ISO 9001:2008
Disposable Single-use
Show More Show Less