Learn More
Corning™ PET Storage Bottles, Square with Cap
Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage
Supplier: Corning™ 431532
Description
Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage. PET resin is lightweight, break-resistant and transparent as glass. Bottles use a linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.
Features:
- Good for storage of media, buffers and other aqueous solutions
- Wide neck provides easier access for liquid transfer
- Screened white enamel graduations are easier to read than molded graduations
- Sterilized to SAL 10−6
- Large white marking spot for easier identification and easy opening linear tear packaging
- Individually bagged
- Validated USP Class VI, noncytotoxic and nonpyrogenic
Chemical Identifiers
| Orange | |
| 37 mm | |
| PET | |
| 12/Pk. | |
| Sterile | |
| Orange | |
| Single-use | |
| Square | |
| GL 45 |
| 1.4 in. | |
| Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage | |
| 24/Cs. | |
| 12 Pk. | |
| GL 45 | |
| ISO 9001:2008 | |
| Yes | |
| HDPE | |
| Plug Seal |
Specifications
| 16.9 oz., 500 mL | |
| 16.9 oz. | |
| 3 in. | |
| 37 mm | |
| PET | |
| 12/Pk. | |
| Sterile | |
| Orange | |
| ISO 9001:2008 | |
| Yes | |
| 176 mm | |
| HDPE | |
| 77 mm |
| 500 mL | |
| Orange | |
| 1.4 in. | |
| Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage | |
| 24/Cs. | |
| 12 Pk. | |
| GL 45 | |
| 100 to 500 mL | |
| Single-use | |
| Square | |
| 6.9 in. | |
| GL 45 | |
| Plug Seal |