missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PET Storage Bottles, Square with Cap

Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage

Supplier:  Corning™ 431531

 View more versions of this product

Catalog No. 07201601


Only null left

Description

Description

Square polyethylene terephthalate (PET) storage bottles are easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage. PET resin is lightweight, break-resistant and transparent as glass. Bottles use a linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.

Features:

  • Good for storage of media, buffers and other aqueous solutions
  • Wide neck provides easier access for liquid transfer
  • Screened white enamel graduations are easier to read than molded graduations
  • Sterilized to SAL 10−6
  • Large white marking spot for easier identification and easy opening linear tear packaging
  • Individually bagged
  • Validated USP Class VI, noncytotoxic and nonpyrogenic


  •  
Specifications

Chemical Identifiers

Orange
37 mm
PET
12/Pk.
Sterile
Orange
Single-use
Square
GL 45
1.4 in.
Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage
24/Cs.
12 Pk.
GL 45
ISO 9001:2008
Yes
HDPE
Plug Seal

Specifications

8.45 oz., 250 mL
8.45 oz.
2.3 in.
37 mm
PET
12/Pk.
Sterile
Orange
ISO 9001:2008
Yes
142.5 mm
HDPE
59 mm
250 mL
Orange
1.4 in.
Easier to handle, require less space (13-20%) on the shelf and are ideal for mixing, sampling and storage
24/Cs.
12 Pk.
GL 45
50 to 250 mL
Single-use
Square
5.6 in.
GL 45
Plug Seal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions