missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Nu-Serum™ IV Growth Serum Replacements
GREENER_CHOICE

Catalog No. CB51004
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CB51004 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CB51004 Supplier Corning™ Supplier No. 355104

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides a low-serum alternative to newborn calf, fetal bovine, and other sera used for cell culture

The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.

  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions

Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.

Specifications

Content And Storage -20°C
Product Type Growth Supplement
Quantity 100 mL