Learn More
Corning™ Microcentrifuge Tubes
Catalog No. 0553870C
Change view
Click to view available options
Capacity (Metric):
1.5 mL
2 mL
Bottom Shape:
Conical
Conical (Self-standing)
Cap Style:
Attached with Silicone O-ring
Unattached
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Bottom Shape Cap Style
0553870C 2 mL Conical (Self-standing) Unattached
0553869C 2 mL Conical (Self-standing) Attached with Silicone O-ring
0553871A 1.5 mL Conical Attached with Silicone O-ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0553870C Supplier Corning™ Supplier No. 430917
Specially designed polypropylene tubes protect sensitive samples and provide added security against leakage and contamination. Corning™ Microcentrifuge Tubes withstand forces up to 20,000 x g.
  • Corning polypropylene microcentrifuge tubes feature screw caps that provide a tight secure seal.
  • Choice of attached cap with silicone O-Ring or unattached rim seal cap.
  • Withstands a maximum 20,000 RCF.
  • Sterile
Specifications
Cap Style Unattached
Closure Color Orange
Closure Material Polypropylene
Capacity (Metric) 2 mL
Color Natural
Material Polypropylene
No. per Pack 50/Pk.
Sterility Sterile
Closure Type Screw Cap
Bottom Shape Conical (Self-standing)
Max. RCF 20,000 x g
Product Type Microcentrifuge Tube
Show More Show Less