missing translation for 'onlineSavingsMsg'
Learn More

Corning™ ITS Premix Universal Culture Supplement
GREENER_CHOICE

Catalog No. p-90935
Change view
Click to view available options
Quantity:
5 mL
20 mL
Formulation:
Lyophilized from a water solution; reconstitute in 20 mL dH2O (stock solution)
Lyophilized from a water solution; reconstitute in 5 mL dH2O (stock solution)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
CB40350 20 mL Lyophilized from a water solution; reconstitute in 20 mL dH2O (stock solution)
CB40351 5 mL Lyophilized from a water solution; reconstitute in 5 mL dH2O (stock solution)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CB40350 Supplier Corning™ Supplier No. 354350
Only null left

Stimulates proliferation of a variety of cells under serum-reduced conditions. Corning™ ITS Premix Universal Culture Supplement contains insulin, human transferrin and selenous acid, the three most universally esential components of defined culture media.

The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.

  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions

Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.

Specifications

Content And Storage 2° to 8°C
Product Type ITS Premix
Sterility Sterile-filtered
Formulation Lyophilized from a water solution; reconstitute in 20 mL dH2O (stock solution)
Quantity 20 mL