missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Gel-Loading Pipet Tips, Flat, 0.2 mm

Corning–Gel-Loading Pipet Tips–Designed with capillary end to allow easy access into vertical and horizontal electrophoresis gels.

$465.21 - $491.13

Specifications

Sterility Non-sterile
Color Natural
Thickness 0.2 mm
Clean Claims DNase, RNase Free
For Use With (Application) Capillary end allows easy access into vertical and horizontal electrophoresis gels, ultra micropipettors
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
07-200-509
View Documents
Corning™
4815
10 μL
Case of 400 Each for $491.13
Only null left
 
07-200-519
View Documents
Corning™
C4884
1 to 200 μL
Case of 400 Each for $465.21
Only null left
 
Description

Description

Corning™ gel-loading polypropylene pipet tips feature a flat 0.2 mm thick capillary end that allows easy access into vertical and horizontal electrophoresis gels. Designed for use with Gilson™ and other popular ultra-micropipettors-200 tips/rack-Nonsterile, RNase-/DNase-free, nonpyrogenic and trace metal-free.
Specifications

Specifications

Non-sterile
0.2 mm
Capillary end allows easy access into vertical and horizontal electrophoresis gels, ultra micropipettors
Rack
Natural
DNase, RNase Free
Gel Loading
Polypropylene
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'