missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Fibronectin, Human
GREENER_CHOICE

Catalog No. CB40008A
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CB40008A 5 mg
CB40008 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CB40008A Supplier Corning™ Supplier No. 356008

Promotes cellular attachment, spreading, and proliferation on tissue-culture surfaces. Corning™ Fibronectin, Human can be used as an additive to serum-free culture media.

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

The principal functions of fibronectin appear to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis.

Specifications

Content And Storage 2° to 8°C
Product Type Fibronectin
pH 11
Quantity 5 mg
Source Human Plasma