Learn More
Corning™ Untreated Culture Dishes
Binds bio-molecules through passive interactions and hydrophobic in nature. Corning™ Untreated Culture Dishes provide an ideal environment for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.
$496.25 - $1246.81
Specifications
| Closure Color | Clear |
|---|---|
| Closure Included | Yes |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Color | Clear |
| Catalog Number | Mfr. No. | Diameter (Metric) | Culture Area | Height (Metric) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) | Culture Area | Height (Metric) | Price | Quantity | |||
|
08-772-25
|
Corning™
430597 |
150 mm | 148 cm2 | 25 mm |
Case of 60 Each for $496.25
|
|
|||
|
08-772-30
|
Corning™
430588 |
35 mm | 8 cm2 | 10 mm |
Case of 500 Each for $1,246.81
|
|
|||
|
08-772-32
|
Corning™
430591 |
100 mm | 55 cm2 | 20 mm |
Case of 500 Each for $1,037.28
|
|
|||
|
08-772-31
|
Corning™
430589 |
60 mm | 21 cm2 | 15 mm |
Case of 500 Each for $663.28
|
|
|||
Description
- Manufactured from optically clear virgin polystyrene
- Not treated for applications where cell attachment is not desired
- Not treated for applications where cell attachment is not desired
- Stacking beads aid in handling
- Vents provide consistent gas exchange
- Sterile
- Sterile
- Nonpyrogenic
Dish style (mm) = actual growth surface diameters: 35 mm dish = 34.4 mm; 60 mm dish = 52.1 mm; 100 mm dish = 83.8 mm; 150 mm dish = 139.1 mm.
Cat. no. 431111, Square dishes with interior bottom dimensions of 224 x 224 mm.
Order Info
For Cell Culture Treated Polystyrene Dishes: Shelf pack 430293 consists of six-pack carriers which hold six sleeves of 10 dishes each–minimizing contamination of unused dishes after sleeve is opened. Supplied in sleeves of 20 dishes (except 150 x 25mm dish which is packaged five per sleeve and 245 x 245mm dish which comes four per sleeve).
Specifications
| Clear | |
| Polystyrene | |
| Clear | |
| Polystyrene | |
| Sterile | |
| Cell Culture Petri Dish |
| Yes | |
| Flat | |
| Single-use | |
| Round | |
| Non-treated | |
| Lid |