Learn More
Corning™ CrystalEX™ 96-Well Protein Crystallization Microplates
Designed for sitting-drop vapor diffusion high throughput protein crystallization
$3,894.36
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
No. of Wells 96
View More Specs
Products 1
Catalog Number Mfr. No. Surface Treatment Price Quantity & Availability  
Catalog Number Mfr. No. Surface Treatment Price Quantity & Availability  
07200790
View Documents Promotion Details
Corning™
C3773
Non-treated
Case of 50 Each for $3,894.36
Only null left
Description
Description
96-well Microplates
  • Features 96 large reservoir (reagent) wells and 96 corresponding protein wells
  • Conical bottom protein wells allow improved centering of the protein drop.
  • Compatible with manual pipettors and automation
  • Novel merged well design provides efficient vapor space for protein crystallization.
  • Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
384-well Microplate
  • Meets ANSI/SBS standard footprint dimensions for microplates
  • Ideal for fully automated crystal screening
  • Features 192 reservoir wells and 192 corresponding protein wells
  • Flat bottom protein wells are optimized for imaging of crystals.
  • Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
Specifications
Specifications
Standard
8 x 12
96
96 flat-bottom reservoir wells and 96 conical-bottom protein wells
50/Cs.
Without Lid
Clear
Non-sterile
50/Cs.
3.1 mm