missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Trace Elements

Catalog No. MT99182CI
Change view
Click to view available options
Description:
Trace Elements A
Trace Elements B
Trace Elements C
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
MT99182CI Trace Elements A
MT99176CI Trace Elements C
MT99175CI Trace Elements B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MT99182CI Supplier Corning™ Supplier No. 25021CI

For Research applications

cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • Trace Elements A includes Cupric Sulfate, Ferric Citrate, Sodium Selenite, and Zinc Sulfate
  • Trace Elements B includes Ammonium Molybdate, Ammonium Vanadate, Manganese Sulfate, Nickel Sulfate, Sodium Silicate, Stannous Chloride and Hydrochloric Acid
  • Trace Elements C includes Aluminum Chloride, Barium Acetate, Cadmium Chloride, Chromic Chloride, Cobalt Dichloride, Germanium Dioxide, Potassium Bromide, Potassium Iodide, Rubidium Chloride, Silver Nitrate, Sodium Fluoride, and Zirconyl Chloride

Specifications

Content And Storage 15° to 30°C
Description Trace Elements A
Form Liquid
Product Type Trace Element
Sterility Sterile-filtered
Format Plastic Bottle
Quantity 100 mL