missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Polysucrose 400, Powder

Catalog No. MT61196RO
Change view
Click to view available options
Quantity:
1 x 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT61196RO 1 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT61196RO Supplier Corning™ Supplier No. 61196RO

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A synthetic polymer of sucrose and epichlorohydrin used for the density gradient separation of cells, cell membranes and virus cells

  • Used to isolate different organelles
  • Nonionic compound

Order Info

Packaged in PET bottles.

Specifications

Content And Storage 15° to 30°C
Form Powder
Product Type Polysucrose 400
Quantity 1 x 500 g