missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Minimum Essential Medium Eagle (Mod.) 1X (MEM) without Glutamine
Guaranteed low endotoxin levels ≤0.025EU/mL.
Supplier: Corning™ 15010CV
Description
Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. This research indicated that additions made to BME aided cell propagation. MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayer, such as fibroblast. This formulation contains sodium bicarbonate and does not contain L-glutamine.Specifications
| Liquid | |
| Sterile | |
| No Glutamine | |
| 7 to 7.4 |
| MEM Eagle/EBSS | |
| Earle's Balanced Salt Solution (EBSS) | |
| 6 x 500 mL | |
| 2° to 8°C |