Learn More
Corning™ Minimum Essential Medium Eagle (Mod.)
Formulated without sodium bicarbonate to prevent clumping due to moisture absorption
Supplier:  Corning™ 50011PB
 View more versions of this product
Catalog No. MT-50-011-PB
Only null left
Explore available promotions
Description
Description
Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.  This research indicated that additions made to BME aided cell propagation.  MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells.  When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts.  This formulation contains Earle's salts and L-glutamine, and does not contain sodium bicarbonate.
Specifications
Specifications
Earle's Balanced Salt Solution (EBSS), Glutamine, Non Essential Amino Acids (NEAA)
No Sodium Bicarbonate
10 L
Product Suggestions
Product Suggestions