missing translation for 'onlineSavingsMsg'
Learn More

Corning™ MEM Vitamin Solution

Catalog No. MT25020CI
Change view
Click to view available options
Quantity:
6 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT25020CI 6 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT25020CI Supplier Corning™ Supplier No. 25020CI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Addition to medium improves growth and viability of propagating cells in culture while reducing the biosynthetic burden on these cells

(This is suggested replacement for 25-020-CIR.)

Specifications

Content And Storage -25° to -15°C
Form Liquid
Product Type MEM Vitamin Solution
Sterility Sterile-filtered
Format Plastic Bottle
Quantity 6 x 100 mL