missing translation for 'onlineSavingsMsg'
Learn More

Corning™ McCoy's 5A (Iwakata and Grace Mod.), 1X

With L-Glutamine

$228.25 - $228.25

Specifications

Product Type McCoy's's 5A (Iwakata & Grace Mod.) 1 X
pH 7 to 7.4
View More Specs

Products 1
Catalog Number Mfr. No. Quantity With Additives Price Quantity  
Catalog Number Mfr. No. Quantity With Additives Price Quantity  
MT10050CV
View Documents
Corning™
10050CV
6 x 500 mL Glutamine
Case of 6 Each for $228.25
Only null left
 
Description

Description

McCoy's 5A medium was originally formulated to meet the culture requirements of Novikoff Hepatoma cells, and has been found to support the indefinite proliferation of Walker 256 carcinoma cells along with a wide range of human and rat normal and transformed cells.  Originally modified by Iwakata and Grace, this version of McCoy's 5A includes increased levels of inositol and glucose.
Specifications

Specifications

McCoy's's 5A (Iwakata & Grace Mod.) 1 X
7 to 7.4
Videos
SDS
Documents

Documents