missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Fetal Bovine Serum, Premium (Low IgG)

Catalog No. MT35073CV
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT35073CV 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT35073CV Supplier Corning™ Supplier No. 35073CV

Low IgG FBS is used in IgG Antibody production, in the propagation of organisms that are normally hindered by the presence of IgGs, or any project in which downstream results would be hindered by exogenous IgGs.

Fetal Bovine Serum (FBS) is a time tested, universal supplement used in cell culture. It provides many compounds that have been shown to satisfy specific metabolic requirements for the culture of cells, including hormones, growth factors, cell attachment factors, pH buffer, protease inhibitors, vitamins, minerals, lipids, and a variety of undefined components. Serum also protects against toxic effects associated with pH change, the presence of heavy metal ions, endotoxin, and proteolytic activity.

Learn more about our custom serum options.

Sera Processing

Using industry leading techniques, blood is aseptically collected at abattoirs, centrifuged to isolate the serum and immediately frozen for testing and further processing. The raw sera is then true pooled and is triple 0.1 micron sterile filtered with a Sterility Assurance Level (SAL) of 10-3. Every lot then must pass USP <71> and EP 2.6.1 sterility testing, growth promotion testing, as well as test negative for mycoplasma and number of industry-monitored viruses. Authenticity of the serum is verified using industry leading measurements of Gamma-Glutamyl Transferase and Corning’s high internal standard of ≤ 10 IU/L. Additional testing includes endotoxin, hemoglobin, pH, osmolality, total protein, and a biochemical profile that contains various metals, proteins, hormones and other metabolites.

EDQM Certificates of Suitability

Fetal Bovine Serum, United States Origin

This product has been granted a Certificate of Suitability, R0-CEP 2018-271, by the European Directorate for the Quality of Medicines (EDQM) certifying that the product meets the criteria described in the current version of the monograph no. 1483 of the European Pharmacopoeia “Product with risk of transmitting agent of animal spongiform encephalopathies.”

Origin Statement

Corning’s premium FBS is only collected from government approved abattoirs in the United States, Australia, and New Zealand. All fetal bovine serum is derived from fetuses collected from healthy bovine cows and heifers that have passed ante and post mortem inspection and were found free of contagious diseases.

Corning’s regular FBS is derived from government registered facilities in North, Central, and South America and outside the United States. All fetal bovine serum is derived from fetuses collected from healthy bovine cows and heifers that have passed ante and post mortem inspection and were found free of contagious diseases. The material was exported from the country of origin accompanied with a Veterinarian Health Certificate. 

'

Specifications

Certifications/Compliance cGMP
Color Amber Yellow to Reddish
Content And Storage -25° to -15°C
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Source Bovine Fetus
Grade Premium
Endotoxin Concentration 8.9EU/ mL
For Use With (Application) Used in IgG Antibody production, in the propagation of organisms that are normally hindered by the presence of IgGs, or any project in which downstream results would be hindered by exogenous IgGs.
Osmolality 305 ± 45mOsm/kg
pH Test and Report
USDA Safety Testing Included USDA Inspected
Show More Show Less