missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Insulin-Transferrin-Selenium (ITS)

Catalog No. MT25800CR
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT25800CR 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT25800CR Supplier Corning™ Supplier No. 25800CR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.

  • 100X solution
  • Animal free formulation
  • Contains 1000mg/L human recombinant insulin, 550mg/L human recombinant transferrin, 0.67mg/L selenious acid
  • Quantity: 10mL

Specifications

Content And Storage 2° to 8°C
Form Liquid
Product Type ITS (Insulin, Transferrin, Selenium)
Sterility Sterile-filtered
Format Plastic Bottle
pH 3 ±0.4
Quantity 10 mL