missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Improved MEM (Richter's Mod.), 1X

With L-Glutamine

$391.70 - $397.23

Specifications

Quantity 6 x 500 mL
pH 7 to 7.4
Content And Storage 2° to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
MT10024CV
View Documents
Corning™
10024CV
Improved MEM
Case of 6 Each for $397.23
Only null left
 
MT10026CV
View Documents
Corning™
10026CV
MEM Richters
Case of 6 Each for $391.70
Only null left
 
Description

Description

A modified formulation of Minimal Essential Medium, Improved Minimum Essential Medium (IMEM) contains a higher concentration of many components used in MEM along with additional amino acids, vitamins, and metabolites.
Specifications

Specifications

6 x 500 mL
7 to 7.4
2° to 8°C
Videos
SDS
Documents

Documents