missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell Culture Media: Cell Culture Supplements

Catalog No. MT46033CI
Change view
Click to view available options
pH:
5.2 to 0.1
Neutral
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
MT46033CI 5.2 to 0.1
MT25037CI Neutral
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT46033CI Supplier Corning™ Supplier No. 46033CI

Used as a nutrient source in cell culture media. Cell Culture Media: Cell Culture Supplements are produced under the highest industry standards to ensure superior results.

  • Certificate of Analysis available

Specifications

Content And Storage 15° to 30°C
Product Type Sodium Acetate
Sterility Sterile-filtered
pH 5.2 to 0.1
Quantity 100 mL