missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Cell-Tak Cell and Tissue Adhesive

Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Specifications
Specifications
| Content And Storage | Stable for at least three months at 2° to 8°C. Do not freeze. |
| Description | Molecular weight: 110 to 114kd |
| Concentration | 1 to 5μg/ cm2 of growth surface depending on cell type |
| Form | Liquid |
| Product Type | Cell and Tissue Adhesive |
| Formulation | As liquid in 5% acetic acid |
| Quantity | 1 mg |
| Source | Polyphenolic Proteins secreted by Mytilus edulis |