missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell-Tak Cell and Tissue Adhesive
GREENER_CHOICE

Catalog No. CB40240
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CB40240 1 mg
CB40241 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CB40240 Supplier Corning™ Supplier No. 354240
Only null left

Attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP Polymer, and biological materials. Corning™ Cell-Tak Cell and Tissue Adhesive is a specially formulated protein solution extracted from marine mussel, Mytilus edulis.

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Specifications

Content And Storage Stable for at least three months at 2° to 8°C. Do not freeze.
Description Molecular weight: 110 to 114kd
Concentration 1 to 5μg/ cm2 of growth surface depending on cell type
Form Liquid
Product Type Cell and Tissue Adhesive
Formulation As liquid in 5% acetic acid
Quantity 1 mg
Source Polyphenolic Proteins secreted by Mytilus edulis