missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Calcein AM Fluorescent Dye
Reliable and consistent dye used for the detection of live cells and key functional activities in variety of cell-based assays
$396.55 - $400.11
Specifications
| Content And Storage | -20°C |
|---|---|
| For Use With (Equipment) | Falcon FluoroBlok™ Cell culture inserts and insert systems to label cells |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
08-774-500
|
Corning™
354216 |
500 μg |
Each for $396.55
|
|
|||||
|
08-774-501
|
Corning™
354217 |
1 mg |
Each for $400.11
|
|
|||||
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Specifications
| -20°C | |
| Falcon FluoroBlok™ Cell culture inserts and insert systems to label cells |
For research use only.