missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Calcein AM Fluorescent Dye

Catalog No. 08774500
Change view
Click to view available options
Quantity:
500 μg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08774500 500 μg
08774501 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08774500 Supplier Corning™ Supplier No. 354216

Reliable and consistent dye used for the detection of live cells and key functional activities in variety of cell-based assays

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Specifications

Content And Storage -20°C
For Use With (Equipment) Falcon FluoroBlok™ Cell culture inserts and insert systems to label cells
Quantity 500 μg

For research use only.