missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Poly-D-Lysine/Laminin Culture Slide
GREENER_CHOICE

Catalog No. CB354688
Change view
Click to view available options
No. of Wells:
8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
CB354688 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CB354688 Supplier Corning™ Supplier No. 354688

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tested for the ability to promote neurite outgrowth with NG108 rat glioma/mouse neuroblastoma cells

Corning™ BioCoatTM 8-well culture slide with a uniform application of PDL/Laminin. Glass slide with polystyrene vessel, lid, and safety removal tool. 4/pack, 12/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.

Shelf life: >= 3 months from date of shipment

Specifications

Type Culture Slide
For Use With (Application) Adhesion, growth and differentiation of neuronal and transfected cell lines
Storage Requirements 2° to 8°C
No. per Pack 3/Pk.
No. per Case 12/Cs.
No. of Wells 8