Learn More
Corning™ BioCoat™ FluoroBlok™ Fibronectin Cell Culture Inserts

Detect migrated cells below membrane using a fluorescence plate reader
Supplier: Corning™ 354597
Description
Packaged 12 inserts in a 24 well plate, 2 plates per case
Corning BioCoat™ cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
- Available as convenient, individual inserts are packaged in ready-to-use 24-well plates and as a one-piece, automation friendly 24-Multiwell format (clear PET membrane only)
- Combined benefits of Falcon™ inserts coated with HFN for enhanced cell attachment and growth
- Individual inserts contains a 3.0 μm pore size PET or Corning™ FluoroBlok PET membrane
Storage: Store at 2-8°C, away from direct sunlight, do not freeze.; Shelf life: At least 3 months from date of shipment
Order Info
Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates
Specifications
| 4.2 cm2 | |
| 3 μm | |
| Promote attachment of BHK-21 Cells in Serum-Free Media | |
| 12/Pk. | |
| 24 |
| PET | |
| Plate | |
| 2° to 8°C | |
| 24/Cs. |