missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Fibronectin Culture Slide, Human
GREENER_CHOICE

Catalog No. 08774285
Change view
Click to view available options
No. of Wells:
4
8
Type:
Chambers and Microslides
Culture Slide
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells Type
08774285 4 Chambers and Microslides
08774214 8 Culture Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08774285 Supplier Corning™ Supplier No. 354559

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells

Human Fibronectin (HFN) is a widely distributed glycoprotein that is used as a substrate to promote attachment of cells through its central-binding domain RGD sequence. HFN is a product of most mesenchymal and epithelial cells and is present in both the ECM and plasma. The principal function of HFN appears to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced-serum culture
  • Cell adhesion assays
  • Studies of effects of HFN on cell behavior
  • Improving survival of primary cells in culture

Source

  • Human plasma

  • NOTE: Source material tested for hepatitis B antigen and HIV-1 antibody

    Quality Control

    • Tested for ability to promote attachment and spreading of BHK-21 hamster kidney cells
    • Tested and found negative for bacteria and fungi
    • Fibronectin purity >90% by SDS-PAGE

    Storage

  • 2°C to 8°C. Do not freeze.
  • Specifications

    Type Chambers and Microslides
    For Use With (Application) Adhesion, growth and differentiation of neuronal and transfected cell lines
    Storage Requirements 2° to 8°C
    No. per Pack 3/Pk.
    No. per Case 12/Cs.
    Surface Coating Fibronectin
    No. of Wells 4