missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Collagen I Inserts with PET Membrane
GREENER_CHOICE

Catalog No. p-2386680
Change view
Click to view available options
Pore Size:
0.4 μm
3 μm
Type:
Well Plate
Quantity:
2
4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Type Quantity
0877419 0.4 μm Well Plate 2
07201235 3 μm Well Plate 4
07201236 3 μm Well Plate 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0877419 Supplier Corning™ Supplier No. 354444
Only null left

Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

  • Each lot is tested for the ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
  • Tested and found negative for bacteria and fungi
  • Cellware is also tested for ability to promote firm attachment of rat cerebellar granule cells
  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

For research use only.

Order Info

Bulk quantities are also available; contact your Fisher Scientific Customer Service Representative.

Specifications

Inserts per Plate 12
Culture Area 0.3 cm2
Material PET
Pore Size 0.4 μm
Type Well Plate
For Use With (Application) Adhesion, growth and differentiation of neuronal and transfected cell lines
Quantity 2
Storage Requirements 2° to 8°C
No. per Pack 12/Pk.
No. per Case 24/Cs.
Surface Coating Collagen
No. of Wells 24
Show More Show Less