missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Collagen I Inserts with PET Membrane
Description
- Each lot is tested for the ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
- Tested and found negative for bacteria and fungi
- Cellware is also tested for ability to promote firm attachment of rat cerebellar granule cells
- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For research use only.
Order Info
Bulk quantities are also available; contact your Fisher Scientific Customer Service Representative.
Specifications
Specifications
| Culture Area | 0.3 cm2 |
| Material | PET |
| Pore Size | 0.4 μm |
| Type | Well Plate |
| For Use With (Application) | Adhesion, growth and differentiation of neuronal and transfected cell lines |
| Quantity | 2 |
| Storage Requirements | 2° to 8°C |
| No. per Pack | 12/Pk. |
| No. per Case | 24/Cs. |
| Surface Coating | Collagen |
| Show More |