missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Collagen I Glass Multiwell Culture Slide
GREENER_CHOICE

Catalog No. p-2386644
Change view
Click to view available options
No. of Wells:
4
8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
08774213 8
08774283 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08774213 Supplier Corning™ Supplier No. 354630
Only null left

Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture

Source

  • Rat tail tendon
  • Quality Control

    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE

    Storage

  • 4°C to 30°C under dry conditions.
  • Specifications

    Type Culture Slide
    For Use With (Application) Adhesion, growth and differentiation of neuronal and transfected cell lines
    No. per Pack 3/Pk.
    No. per Case 12/Cs.
    Surface Coating Collagen I
    No. of Wells 8