missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Assay System
Description
Includes
24-Multiwell™ Insert plate, specialized media to perform 24 assays, feeder tray, and lid.
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
Specifications
| Closure Included | Yes |
| Color | Clear |
| Culture Area | 0.3 cm2 |
| Material | PET |
| Sterility | Non-sterile |
| Surface Treatment | Collagen |
| Type | Cell Culture Insert System |
| Well Count | 24 |
| For Use With (Application) | Rapid differentiation of Caco 2 cells in vitro for robotic screening of prospective pharmaceuticals for oral bioavailability and absorption |
| Includes | 24 multiwell insert plate, specialized media to perform 24 assays, feeder tray and lid |
| Show More |