Learn More
Corning™ BioCoat™ #1 German Glass Coverslips, Round
Coated with a variety of extracellular matrix substrates. Corning™ BioiCoat™ No. 1 German Glass Coverslips provide an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.
$484.23 - $721.07
Specifications
| Type | Slide Coverslip |
|---|---|
| Material | No. 1 German Glass |
| For Use With (Application) | Fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy, phase contrast microscopy |
| Catalog Number | Mfr. No. | Diameter (Metric) | Surface Treatment | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) | Surface Treatment | Price | Quantity | ||||
|
08-774-383
|
Corning™
354085 |
12 mm | Poly-L Lysine |
Case of 80 Each for $701.02
|
|
||||
|
08-774-384
|
Corning™
354086 |
12 mm | Poly-D Lysine |
Case of 80 Each for $651.15
|
|
||||
|
08-774-386
|
Corning™
354088 |
22 mm | Fibronectin |
Case of 60 Each for $597.10
|
|
||||
|
08-774-385
|
Corning™
354087 |
12 mm | Laminin/Poly-D Lysine |
Case of 80 Each for $721.07
|
|
||||
|
08-774-387
|
Corning™
354089 |
22 mm | Collagen Type I |
Case of 60 Each for $484.23
|
|
||||
Description
Corning™ BioCoat™ German Glass coverslips with surface treatment (see individual Specification tables for coating). Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
Shelf life: ≥ 3 months from date of shipment
Specifications
| Slide Coverslip | |
| No. 1 German Glass | |
| Fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy, phase contrast microscopy |