Learn More
Corning™ BioCoat™ #1 German Glass Coverslips, Round
Coated with a variety of extracellular matrix substrates. Corning™ BioiCoat™ No. 1 German Glass Coverslips provide an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.
Supplier: Corning™ 354087
Description
Corning™ BioCoat™ German Glass coverslips with surface treatment (see individual Specification tables for coating). Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
Shelf life: ≥ 3 months from date of shipment
Specifications
| Slide Coverslip | |
| No. 1 German Glass | |
| Fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy, phase contrast microscopy | |
| 80 |
| Laminin/Poly-D Lysine | |
| 0.472 in. | |
| 12 mm | |
| 80 |