Learn More
Corning™ 96-Well Next Generation CrystalEX Protein Crystallization Microplates
Optimized for high-throughput sitting-drop protein crystallization
$2,169.67 - $3,947.84
Specifications
Barcode Alphanumeric
Lid Without Lid
Array 8 x 12
Color Clear
No. of Wells 96
View More Specs
Products 3
Catalog Number Mfr. No. Well Shape Volume (Metric) Well Material Price Quantity & Availability  
Catalog Number Mfr. No. Well Shape Volume (Metric) Well Material Price Quantity & Availability  
07201837
View Documents Promotion Details
Corning™
C3550
Conical Flat 1 μL Polymer
Case of 50 Each for $2,169.67
Only null left
07201831
View Documents Promotion Details
Corning™
C3552
Round, Flat Bottom (F-well) 2 μL PZero
Case of 50 Each for $3,688.64
Only null left
07201832
View Documents Promotion Details
Corning™
3553
Conical Flat 2 μL PZero
Case of 50 Each for $3,947.84
Only null left
Description
Description
  • Corning™ 96-well and 384-well crystallization microplates are optimized for high throughput protein crystal growth and screening
  • Designed for sitting drop applications
  • Meets ANSI/SBS standard footprint dimensions for microplates
  • Ideal for fully automated crystal screening
  • Multiple formats and versatility for custom options to maximize crystal formation, identification and analysis, and harvesting
  • - Choose from four unique protein well shapes
    - Options include one or three protein wells per reservoir well
  • Cyclic olefin copolymer (COC) offers strong chemical compatibility and good optical clarity
  • Reservoir numbers are embossed on each individual well for easy identification
Specifications
Specifications
Alphanumeric
8 x 12
96
Non-treated
10/Pk.
Without Lid
Clear
Non-sterile
50/Cs.