Learn More
Corning™ 96-Well Next Generation CrystalEX Protein Crystallization Microplates
Catalog No. 07201831
Change view
Click to view available options
Well Shape:
Conical Flat
Round, Flat Bottom (F-well)
Volume (Metric) Well:
1 μL
2 μL
Material:
Polymer
PZero
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape Volume (Metric) Well Material
07201831 Round, Flat Bottom (F-well) 2 μL PZero
07201837 Conical Flat 1 μL Polymer
07201832 Conical Flat 2 μL PZero
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07201831 Supplier Corning™ Supplier No. C3552
Only null left
Optimized for high-throughput sitting-drop protein crystallization
  • Corning™ 96-well and 384-well crystallization microplates are optimized for high throughput protein crystal growth and screening
  • Designed for sitting drop applications
  • Meets ANSI/SBS standard footprint dimensions for microplates
  • Ideal for fully automated crystal screening
  • Multiple formats and versatility for custom options to maximize crystal formation, identification and analysis, and harvesting
  • - Choose from four unique protein well shapes
    - Options include one or three protein wells per reservoir well
  • Cyclic olefin copolymer (COC) offers strong chemical compatibility and good optical clarity
  • Reservoir numbers are embossed on each individual well for easy identification
Specifications
Barcode Alphanumeric
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material PZero
Volume (Metric) Well 2 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round, Flat Bottom (F-well)
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Show More Show Less