missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Clear-Bottom Nonbinding Surface (NBS™) Microplates

Enhanced signal-to-noise ratio
$4,068.54 - $4,170.38
Specifications
| Format | Standard |
|---|---|
| Lid | Without Lid |
| Array | 8 x 12 |
| For Use With (Equipment) | Plate readers, liquid handlers and robotic systems |
| Material | Polystyrene |
Description
Specifications
| Standard | |
| 8 x 12 | |
| Polystyrene | |
| 75 to 200 μL | |
| Non-binding | |
| For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays | |
| 25/Pk. | |
| 39°F to 99°F | |
| Clear |
| Without Lid | |
| Plate readers, liquid handlers and robotic systems | |
| 96 | |
| Non-sterile | |
| Flat Bottom (F-well) | |
| 100/Cs. | |
| 100/Cs. | |
| 4°C to 37°C |