missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Clear-Bottom Nonbinding Surface (NBS™) Microplates
GREENER_CHOICE

Catalog No. 07200770
Change view
Click to view available options
Color:
Black, Clear Bottom
White, Clear Bottom
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07200770 Black, Clear Bottom
07200732 White, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07200770 Supplier Corning™ Supplier No. 3651
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enhanced signal-to-noise ratio

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • Specifications

    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color Black, Clear Bottom
    For Use With (Equipment) Plate readers, liquid handlers and robotic systems
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment Non-binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Temperature Range (English) 39°F to 99°F
    Temperature Range (Metric) 4°C to 37°C
    Well Color Clear
    Show More Show Less