missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Untreated 245mm Square BioAssay Dishes
Designed with a generous growth area of 500cm2 to allow for larger assays and better overall use of space in incubators. Corning™ Untreated 245mm Square Low Profile BioAssay Dishes provide a superior option for applications where cell attachment is not desired.
Supplier: Corning™ 431111
Description
Improve the efficiency of microbial assays including library replication, colony picking, and plaque lift studies.
- Polystyrene dishes feature stacking beads for stable stacking
- Compatible with various automated colony picking and gridding robots
- Supplied with vented lid to provide consistent gas exchange
- Sterile, certified nonpyrogenic
- Patented (U.S. Patent No.. 5,856,176) design allows for easier handling
Specifications
| Clear | |
| Polystyrene | |
| 150 mL (Well) | |
| 500 sq. cm | |
| Polystyrene | |
| Sterile | |
| Lid, Handles | |
| 245 x 245 x 25 mm | |
| 245 mm | |
| 18 mm |
| Closure Included | |
| Square | |
| Clear | |
| Single-use | |
| Square | |
| Cell Culture Dish | |
| 245 x 245 mm | |
| 245 mm | |
| 25 mm |