missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 0.25% Trypsin in HBSS w/o Calcium and Magnesium

Catalog No. MT25050CI
Change view
Click to view available options
Quantity:
6 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT25050CI 6 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT25050CI Supplier Corning™ Supplier No. 25050CI

Enzymatic dissociation reagents tested for porcine parvovirus

Specifications

Content And Storage -20°C
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile
Without Additives No Calcium, No Magnesium
pH 7 to 7.6
Quantity 6 x 100 mL