missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 0.25% Trypsin, 0.1% EDTA in HBSS w/o Calcium, Magnesium and Sodium Bicarbonate

Catalog No. MT25053CI
Change view
Click to view available options
Quantity:
6 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT25053CI 6 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT25053CI Supplier Corning™ Supplier No. 25053CI
  • ONLINE EXCLUSIVE PRICE
    $163.26
    $100.33 / Case of 6 Each

    Save $62.93

    39.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used to enzymatically release adherent cells from tissue culture plates for passaging.

  • EDTA enhances efficacy of trypsin
  • Manufactured under industry-leading standards for low endotoxin content and superior results
  • Certificate of Analysis available
  • Enzymatic dissociation reagents tested for porcine parvovirus

Specifications

Content And Storage -20°C
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile
Without Additives No Calcium, No Magnesium, No Sodium Bicarbonate
pH 7 to 7.6
Quantity 6 x 100 mL