Learn More
Corn Oil, NF, Spectrum™ Chemical
CO136, 8001-30-7
Supplier:  Spectrum Chemical Mfg Cor CO136500MLGL
 View more versions of this product
Catalog No. 18601849
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Corn Oil, NF its uses include as a solvent vehicle for topical preparations and as a glidant excipient for oral dosage tablets and capsules. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
8001-30-7
Amber Glass Bottle
NF
100%
500 mL
Chemical Identifiers
8001-30-7
Product Suggestions
Product Suggestions