missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corn Oil, MP Biomedicals
Oleic Acid: 43-47%; Linoleic Acid: 39-42%; Palmitic Acid: 7-8%; Stearic Acid: 3-4%, Arachidic Acid: 0.5%; Approx. 7.6 lbs/gal
$74.36 - $365.31
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN90141401
|
MP Biomedicals Inc
0290141401 |
1 lb. |
Each for $74.36
|
|
|||||
|
ICN90141405
|
MP Biomedicals Inc
0290141405 |
5 lb. |
Each for $94.79
|
|
|||||
|
ICN90141425
|
MP Biomedicals Inc
0290141425 |
25 lb. |
Each for $365.31
|
|
|||||
Chemical Identifiers
| 8001-30-7 |
Specifications
| 8001-30-7 | |
| 1 lb. | |
| Corn Oil |
Safety and Handling
Recommended Storage : Room Temperature