missing translation for 'onlineSavingsMsg'
Learn More

Corn Oil, MP Biomedicals

Catalog No. ICN90141401
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN90141401 1 lb.
ICN90141405 5 lb.
ICN90141425 25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN90141401 Supplier MP Biomedicals Inc Supplier No. 0290141401
Only null left

Oleic Acid: 43-47%; Linoleic Acid: 39-42%; Palmitic Acid: 7-8%; Stearic Acid: 3-4%, Arachidic Acid: 0.5%; Approx. 7.6 lbs/gal

Chemical Identifiers

CAS 8001-30-7

Specifications

Quantity 1 lb.
Chemical Name or Material Corn Oil
Recommended Storage Room Temperature