missing translation for 'onlineSavingsMsg'
Learn More

(-)-Corey Lactone 98.0+%, TCI America™

Catalog No. H1418200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1418200MG 200 mg
H14181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1418200MG Supplier TCI America Supplier No. H1418200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32233-40-2
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00075234
InChI Key VYTZWRCSPHQSFX-GBNDHIKLSA-N
Synonym --corey lactone diol, 3ar,4s,5r,6as-5-hydroxy-4-hydroxymethyl hexahydro-2h-cyclopenta b furan-2-one, --corey lactone, corey lactone diol, --corey diol corey lactone, 3ar,4s,5r,6as---hexahydro-5-hydroxy-4-hydroxymethyl-2h-cyclopenta b furan-2-one, 1s,6s,5r,7r-7-hydroxy-6-hydroxymethyl-2-oxabicyclo 3.3.0 octan-3-one, 3ar,4s,5r,6as-hexahydro-5-hydroxy-4-hydroxymethyl-2h-cyclopenta b furan-2-one, 3ar,4s,5r,6as-5-hydroxy-4-hydroxymethyl-3,3a,4,5,6,6a-hexahydrocyclopenta b furan-2-one, l-corey lactone
PubChem CID 2724453
IUPAC Name (3aR,4S,5R,6aS)-5-hydroxy-4-(hydroxymethyl)-hexahydro-2H-cyclopenta[b]furan-2-one
SMILES OC[C@H]1[C@H](O)C[C@@H]2OC(=O)C[C@H]12

Specifications

Melting Point 116°C
Color White
Quantity 200 mg
Formula Weight 172.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Corey Lactone