accessibility menu, dialog, popup

UserName

CAS RN 32233-40-2

IUPAC Name: (3aR,4S,5R,6aS)-5-hydroxy-4-(hydroxymethyl)-hexahydro-2H-cyclopenta[b]furan-2-one
Synonyms: (3aR,4S,5R,6aS)-5-hydroxy-4-(hydroxymethyl)-hexahydrocyclopenta[b]furan-2-one
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: VYTZWRCSPHQSFX-GBNDHIKLSA-N
SMILES: OC[C@H]1[C@H](O)C[C@@H]2OC(=O)C[C@H]12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)