missing translation for 'onlineSavingsMsg'
Learn More

(-)-Corey Lactone 4-Phenylbenzoate 98.0+%, TCI America™

Catalog No. p-7125634
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2756200MG 200 mg
C27561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2756200MG Supplier TCI America Supplier No. C2756200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31752-99-5
Molecular Formula C21H20O5
Molecular Weight (g/mol) 352.39
MDL Number MFCD00078077
InChI Key SZJVIFMPKWMGSX-AKHDSKFASA-N
Synonym corey lactone 4-phenylbenzoate, --corey lactone 4-phenylbenzoate alcohol, 3ar,4s,5r,6as-4-hydroxymethyl-2-oxohexahydro-2h-cyclopenta b furan-5-yl biphenyl-4-carboxylate, --corey lactone 5-4-phenylbenzoate, --corey lactone 4-phenylbenzoate, --corey lactone, 4-phenylbenzoate alcohol, corey lactone p-phenylbenzoate alcohol, 3ar,4s,5r,6as-1,1'-biphenyl-4-carboxylic acid, hexahydro-4-methoxymethyl-2-oxo-2h-cyclopentafuran-5-yl ester, 3ar,4s,5r,6as-4-hydroxymethyl-2-oxohexahydro-2h-cyclopenta b furan-5-yl 1,1'-biphenyl-4-carboxylate, 3ar,4s,5r,6as-4-hydroxymethyl-2-oxo-3,3a,4,5,6,6a-hexahydrocyclopenta b furan-5-yl 4-phenylbenzoate
PubChem CID 2724295
IUPAC Name (3aR,4S,5R,6aS)-4-(hydroxymethyl)-2-oxo-hexahydro-2H-cyclopenta[b]furan-5-yl [1,1'-biphenyl]-4-carboxylate
SMILES OC[C@H]1[C@@H](C[C@@H]2OC(=O)C[C@H]12)OC(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 200 mg
Formula Weight 352.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Corey Lactone 4-Phenylbenzoate
TSCA No
Recommended Storage Refrigerator