missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COQ9 Monoclonal Antibody (K94030_7D7), Invitrogen™

Catalog No. MA556244
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556244 200 μL
MA555864 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556244 Supplier Invitrogen™ Supplier No. MA556244
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: QHSETQGAEK PDPESSHSPP RYTDQGGEEE EDYESEEQLQ HRILTAALEF VPAHGWTAEA IAEGAQSLGL SSAAASMFGK DGSELILHFV TQCNTRLTRV LEEEQKLVQL GQAEKRKTDQ

Coenzyme Q10 (CoQ10), or ubiquinone, is a mobile lipophilic electron carrier critical for electron transfer by the mitochondrial inner membrane respiratory chain. COQ9 is 1 of several enzymes involved in biosynthesis of CoQ10 and likely functions in modification of the benzoquinone ring.

Specifications

Antigen COQ9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone K94030_7D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene COQ9
Gene Accession No. O75208
Gene Alias 2310005O14Rik; C16orf49; C78387; coenzyme Q9; coenzyme Q9 homolog; coenzyme Q9 homolog (yeast); COQ10D5; COQ9; HSPC326; PSEC0129; Ubiquinone biosynthesis protein COQ9, mitochondrial
Gene Symbols COQ9
Host Species Mouse
Immunogen Recombinant human COQ9 (immunogen range is 61-180).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57017
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less