missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COQ6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113549
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113549 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113549 Supplier Invitrogen™ Supplier No. PA5113549
Only null left

Rabbit Polyclonal Antibody

COQ6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

COQ6 (Coenzyme Q10 monooxygenase 6) is also named as CGI-10 and belongs to the ubiH/COQ6 family. It is an evolutionarily conserved flavin-dependent monooxygenase required for biosynthesis of coenzyme Q10, or ubiquinone. It has 3 isoforms with the molecular mass of 51 kDa, 43 kDa and 49 kDa. In inner ear, Coq6 is expressed in spiral ganglion, stria vascularis cells, and spiral ligament. Defects in COQ6 are the cause of coenzyme Q10 deficiency, primary, type 6 (COQ10D6).

Specifications

Antigen COQ6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Coq6
Gene Accession No. Q8R1S0, Q9Y2Z9
Gene Alias 5930427M12Rik; AW742344; CGI10; CGI-10; Coenzyme Q10 monooxygenase 6; coenzyme Q6 homolog (yeast); coenzyme Q6 homolog, monooxygenase; coenzyme Q6 monooxygenase; coenzyme Q6, monooxygenase; COQ10D6; Coq6; ubiquinone biosynthesis monooxygenase COQ6; ubiquinone biosynthesis monooxygenase COQ6, mitochondrial
Gene Symbols Coq6
Host Species Rabbit
Immunogen Recombinant Human Ubiquinone biosynthesis monooxygenase COQ6, mitochondrial protein (208-339AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217707, 51004
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less