missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COPS3 Recombinant Rabbit Monoclonal Antibody (23GB3780)
GREENER_CHOICE

Catalog No. MA553642
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553642 100 μL
MA553643 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553642 Supplier Invitrogen™ Supplier No. MA553642
Only null left

Rabbit Recombinant Monoclonal Antibody

COPS3 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The protein encoded by this gene possesses kinase activity that phosphorylates regulators involved in signal transduction. It phosphorylates I kappa-Balpha, p105, and c-Jun. It acts as a docking site for complex-mediated phosphorylation. The gene is located within the Smith-Magenis syndrome region on chromosome 17.

Specifications

Antigen COPS3
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3780
Concentration 0.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cops3
Gene Accession No. O88543, Q68FW9, Q9UNS2
Gene Alias COP9 (constitutive photomorphogenic) homolog, subunit 3; COP9 (constitutive photomorphogenic) homolog, subunit 3 (Arabidopsis thaliana); COP9 (constitutive photomorphogenic), subunit 3; COP9 complex S3; COP9 complex subunit 3; COP9 constitutive photomorphogenic homolog subunit 3; COP9 signalosome complex subunit 3; COP9 signalosome subunit 3; COPS 3; COPS3; Csn3; JAB1-containing signalosome subunit 3; SGN3; signalosome subunit 3
Gene Symbols Cops3
Host Species Rabbit
Immunogen A synthetic peptide of human COPS3 (370-423AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26572, 287367, 8533
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less