missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Sulfate, Pentahydrate, Granular, FCC, 98-102%, Spectrum™ Chemical
C1431, 7758-99-8, CuSO4·5H2O
$469.43 - $9624.47
Chemical Identifiers
| CAS | 7758-99-8 |
|---|---|
| Molecular Formula | CuH10O9S |
| Molecular Weight (g/mol) | 249.68 |
| MDL Number | MFCD00149681 |
| InChI Key | JZCCFEFSEZPSOG-UHFFFAOYSA-L |
| IUPAC Name | copper(2+) pentahydrate sulfate |
| SMILES | O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18601842
|
Spectrum Chemical Mfg Cor
C1431125GM |
125 g | Poly Bottle |
Each for $469.43
|
|
||||
|
18601843
|
Spectrum Chemical Mfg Cor
C1431500GM |
500 g | Poly Bottle |
Each for $895.97
|
|
||||
|
18601844
|
Spectrum Chemical Mfg Cor
C143125KG |
2.5 kg | Poly Pail |
Each for $2,610.10
|
|
||||
|
18601845
|
Spectrum Chemical Mfg Cor
C143112KG |
12 kg | Poly Pail |
Each for $6,041.03
|
|
||||
|
18601846
|
Spectrum Chemical Mfg Cor
C143150KGBL |
50 kg | Fiber Drum |
Each for $9,624.47
|
|
||||
Description
Spectrum™ Chemical Copper Sulfate, Pentahydrate, Granular, FCC is used as a fungicide, algaecide and feed supplement. Find a high quality inventory of Copper Copper Sulfate, Pentahydrate, Granular, FCC. Approved by the Food Chemical Codex, it is used as a fungicide, algaecide, and feed supplement, and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 7758-99-8 | |
| 249.68 | |
| JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
| O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O |
| CuH10O9S | |
| MFCD00149681 | |
| copper(2+) pentahydrate sulfate |
Specifications
| 7758-99-8 | |
| CuH10O9S | |
| UN3077 | |
| JZCCFEFSEZPSOG-UHFFFAOYSA-L | |
| copper(2+) pentahydrate sulfate | |
| 98 to 102% | |
| Poly Bottle |
| 100% | |
| MFCD00149681 | |
| 125 g | |
| O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O | |
| 249.68 | |
| FCC |